Assay Details
Target Gene Details
Entrez Gene ID: | 73102 |
Gene Name: | solute carrier family 22, member 23 |
Gene Aliases: |
3110004L20Rik |
Location: |
Chr.13:34179158-34345487 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a23 | NM_001033167.3 | 4 | 1332 | NP_001028339.1 |
| AK147397.1 | 4 | 1333 | BAE27885.1 | |
| BC053705.1 | 2 | 120 | AAH53705.1 |
More Information
Set Membership: |
|

Genomic Map