Assay Details
Target Gene Details
Entrez Gene ID: | 218397 |
Gene Name: | RAS p21 protein activator 1 |
Gene Aliases: |
Gap, RasGAP, Rasa |
Location: |
Chr.13:85214699-85289493 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasa1 | NM_145452.3 | NP_663427.2 | ||
| AK133354.1 | BAE21614.1 | |||
| AK161497.1 | BAE36423.1 | |||
| BC013637.1 | AAH13637.1 |
More Information
Set Membership: |
|

Genomic Map