Assay Details
Target Gene Details
Entrez Gene ID: | 114304 |
Gene Name: | solute carrier family 28 (sodium-coupled nucleoside transporter), member 3 |
Gene Aliases: |
Cnt3 |
Location: |
Chr.13:58553307-58610877 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc28a3 | NM_022317.3 | 6 | 676 | NP_071712.3 |
| AF305211.1 | 6 | 609 | AAG22552.1 | |
| AK052784.1 | 6 | 677 | BAC35145.1 | |
| AK085680.1 | ||||
| AK145077.1 | 6 | 677 | BAE26226.1 | |
| BC010472.1 | 6 | 664 | AAH10472.1 | |
| BY736278.1 |
More Information
Set Membership: |
|

Genomic Map