Assay Details
Target Gene Details
Entrez Gene ID: | 21984 |
Gene Name: | trophoblast specific protein alpha |
Gene Aliases: |
4311, AV033951, Tb-1, Tb1, Tpbp, b2b1247Clo |
Location: |
Chr.13:60938692-60941935 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpbpa | NM_009411.4 | 3 | 249 | NP_033437.2 |
AK005406.1 | 3 | 249 | BAB24006.1 | |
AK005417.1 | 3 | 250 | BAB24012.1 | |
AK005438.1 | 3 | 249 | BAB24032.1 | |
AK005446.1 | 3 | 250 | BAB24039.1 | |
AK005454.1 | 3 | 251 | BAB24046.1 | |
AK005499.1 | 3 | 250 | BAB24082.1 | |
AK011401.1 | 3 | 250 | BAB27593.1 | |
AK014443.1 | 3 | 251 | BAB29354.1 | |
AK018797.1 | 3 | 250 | BAB31419.1 | |
AK018800.1 | 3 | 249 | BAB31422.1 | |
AK018802.1 | 3 | 249 | BAB31424.1 | |
AK018803.1 | 3 | 250 | BAB31425.1 | |
AK018806.1 | 3 | 250 | BAB31428.1 | |
AK018807.1 | 3 | 250 | BAB31429.1 | |
AK018811.1 | 3 | 250 | BAB31433.1 | |
AK018812.1 | 3 | 250 | BAB31434.1 | |
AK018813.1 | 3 | 250 | BAB31435.1 | |
AK018814.1 | 3 | 250 | BAB31436.1 | |
AK018815.1 | 3 | 249 | BAB31437.1 | |
AK018819.1 | 3 | 250 | BAB31441.1 | |
AK018821.1 | 3 | 250 | BAB31443.1 | |
AK018822.1 | 3 | 251 | BAB31444.1 | |
AK018823.1 | 3 | 250 | BAB31445.1 | |
AK018825.1 | 3 | 251 | BAB31447.1 | |
AK018826.1 | 3 | 250 | BAB31448.1 | |
AK028071.1 | 3 | 249 | BAC25736.1 | |
AK160140.1 | 3 | 243 | BAE35652.1 | |
AY034575.1 | 3 | 225 | AAK58451.1 | |
BC099391.1 | 3 | 236 | AAH99391.1 | |
X17071.1 | 3 | 241 | CAA34917.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
