Assay Details
Target Gene Details
Entrez Gene ID: | 22017 |
Gene Name: | thiopurine methyltransferase |
Gene Aliases: |
AW106912 |
Location: |
Chr.13:47021987-47043338 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpmt | NM_016785.2 | NP_058065.2 | ||
| AF037043.1 | AAD02092.1 | |||
| AF037044.1 | AAD02093.1 | |||
| AF037045.1 | AAD02094.1 | |||
| AF046887.1 | AAC25919.1 | |||
| AK002335.1 | ||||
| BC021598.1 | AAH21598.1 | |||
| CF106558.1 |
More Information
Set Membership: |
|

Genomic Map