Assay Details
Target Gene Details
Entrez Gene ID: | 71988 |
Gene Name: | establishment of cohesion 1 homolog 2 (S. cerevisiae) |
Gene Aliases: |
2410004I17Rik, D030072L07Rik |
Location: |
Chr.14:65819027-65833969 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esco2 | NM_028039.2 | 9 | 1602 | NP_082315.3 |
| AA881675.1 | 1 | 84 | ||
| AK010391.1 | 4 | 427 | BAB26905.1 | |
| AK144685.1 | 9 | 1558 | BAE26012.1 | |
| BC033303.1 | 9 | 1517 | AAH33303.2 | |
| BC071275.1 | 9 | 1535 | AAH71275.1 |
More Information
Set Membership: |
|

Genomic Map