Assay Details
Target Gene Details
Entrez Gene ID: | 13885 |
Gene Name: | esterase D/formylglutathione hydrolase |
Gene Aliases: |
Es-10, Es10, FGH, sid478 |
Location: |
Chr.14:74732297-74750765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esd | NM_001285423.1 | NP_001272352.1 | ||
| NM_016903.5 | NP_058599.1 | |||
| AB025408.1 | BAA84693.1 | |||
| AK007527.1 | BAB25090.1 | |||
| AK010683.1 | BAB27115.1 | |||
| AK021209.1 | BAB32330.1 | |||
| AK079131.1 | BAC37555.1 | |||
| AK088115.1 | BAC40154.1 | |||
| AK163074.1 | ||||
| BC046766.1 | AAH46766.2 |
More Information
Set Membership: |
|

Genomic Map