Assay Details
Target Gene Details
Entrez Gene ID: | 30877 |
Gene Name: | guanine nucleotide binding protein-like 3 (nucleolar) |
Gene Aliases: |
Ns |
Location: |
Chr.14:31012433-31019131 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnl3 | NM_153547.6 | 12 | 1436 | NP_705775.2 |
| NR_104298.1 | 12 | 1388 | ||
| AK077523.1 | 12 | 1367 | BAC36844.1 | |
| AK167183.1 | 12 | 1320 | BAE39318.1 | |
| AY181025.1 | 12 | 1294 | AAO19472.1 | |
| AY185498.1 | 12 | 1294 | AAO19473.1 | |
| BC037996.1 | 12 | 1296 | AAH37996.2 |

Genomic Map