Assay Details
Target Gene Details
Entrez Gene ID: | 114874 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
Mir5131, PA-PLA1 |
Location: |
Chr.14:45593171-45658143 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddhd1 | NM_001039106.3 | 4 | 1301 | NP_001034195.2 |
| NM_001042719.2 | 5 | 1403 | NP_001036184.1 | |
| NM_001284399.1 | 6 | 1375 | NP_001271328.1 | |
| NM_176845.5 | 4 | 1301 | NP_789815.3 | |
| AK014975.1 | 2 | 151 | ||
| AK048690.1 | 5 | 861 | ||
| AK129425.1 | 2 | 712 | BAC98235.1 | |
| AK155294.1 | 6 | 1332 | ||
| AK170271.1 | 5 | 1015 | BAE41676.1 | |
| BC043475.1 | 3 | 555 | AAH43475.1 | |
| CF532335.1 | 3 | 223 | ||
| DQ272481.1 | 4 | 1300 | ABB76652.1 | |
| DQ272482.1 | 4 | 1304 | ABB76653.1 |
More Information
Set Membership: |
|

Genomic Map