Assay Details
Target Gene Details
Entrez Gene ID: | 71000 |
Gene Name: | RIKEN cDNA 4931440J10 gene |
Gene Aliases: |
- |
Location: |
Chr.14:62760501-62830126 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931440J10Rik | NR_045503.1 | |||
| AK016511.1 |
More Information
Set Membership: |
|

Genomic Map