Assay Details
Target Gene Details
Entrez Gene ID: | 16427 |
Gene Name: | inter alpha-trypsin inhibitor, heavy chain 4 |
Gene Aliases: |
ITI-HC4, Itih-4 |
Location: |
Chr.14:30886476-30901986 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itih4 | NM_001159299.2 | 9 | 1162 | NP_001152771.1 |
| NM_001289632.1 | 9 | 1162 | NP_001276561.1 | |
| NM_001289633.1 | 9 | 1162 | NP_001276562.1 | |
| NM_018746.4 | 9 | 1162 | NP_061216.2 | |
| AF023919.1 | 9 | 1125 | AAC25786.1 | |
| AK004862.1 | 9 | 1137 | ||
| AK004893.1 | 9 | 1137 | BAB23649.1 | |
| AK050016.1 | 9 | 1140 | BAC34032.1 | |
| AK050270.1 | 9 | 1137 | BAC34155.1 | |
| BC016500.1 | 9 | 1125 | AAH16500.1 | |
| BC092258.1 | 9 | 1114 | AAH92258.1 | |
| BC094457.1 | 9 | 1131 | AAH94457.1 |
More Information
Set Membership: |
|

Genomic Map