Assay Details
Target Gene Details
Entrez Gene ID: | 22070 |
Gene Name: | tumor protein, translationally-controlled 1 |
Gene Aliases: |
TCTP, Trt, p21, p23 |
Location: |
Chr.14:75845256-75848303 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpt1 | NM_009429.3 | NP_033455.1 | ||
| CJ063333.1 |
More Information
Set Membership: |
|

Genomic Map