Assay Details
Target Gene Details
Entrez Gene ID: | 71412 |
Gene Name: | dehydrogenase/reductase member 2 |
Gene Aliases: |
5430405K24Rik, Hep27 |
Location: |
Chr.14:55124154-55241440 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs2 | NM_027790.2 | NP_082066.2 | ||
| AK017274.1 | BAB30665.1 | |||
| BC117731.2 |
More Information
Set Membership: |
|

Genomic Map