Assay Details
Target Gene Details
Entrez Gene ID: | 56419 |
Gene Name: | diaphanous homolog 3 (Drosophila) |
Gene Aliases: |
4930417P13Rik, Dia2, Diaph3, Drf3, mDia2, mKIAA4117, p134MDia2 |
Location: |
Chr.14:86655361-87141248 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Diap3 | NM_019670.1 | NP_062644.1 | ||
| AF094519.1 | AAC71771.1 | |||
| AK137073.1 | BAE23229.1 | |||
| AK138694.1 | BAE23751.1 | |||
| AK161737.1 | BAE36554.1 | |||
| AK166882.1 | ||||
| AK220493.1 | BAD90291.1 | |||
| BC028920.1 | AAH28920.1 | |||
| BC085191.1 | ||||
| BC086779.1 | AAH86779.1 |

Genomic Map