Assay Details
Target Gene Details
Entrez Gene ID: | 16554 |
Gene Name: | kinesin family member 13B |
Gene Aliases: |
5330429L19Rik, 6030414C01, AI429803, C130021D12Rik, GAKIN |
Location: |
Chr.14:64652490-64809618 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif13b | NM_001081177.1 | NP_001074646.1 | ||
| AK020905.1 |
More Information
Set Membership: |
|

Genomic Map