Assay Details
Target Gene Details
Entrez Gene ID: | 14943 |
Gene Name: | granzyme F |
Gene Aliases: |
CCP4, Ctla-7, Ctla7, MCSP-3 |
Location: |
Chr.14:56205263-56211407 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzmf | NM_010374.3 | NP_034504.1 | ||
| BC100311.1 | AAI00312.1 | |||
| BC145435.1 | ||||
| J03257.1 | AAA37738.1 | |||
| X12823.1 | CAA31310.1 |
More Information
Set Membership: |
|

Genomic Map