Assay Details
Target Gene Details
Entrez Gene ID: | 71988 |
Gene Name: | establishment of cohesion 1 homolog 2 (S. cerevisiae) |
Gene Aliases: |
2410004I17Rik, D030072L07Rik |
Location: |
Chr.14:65819027-65833969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esco2 | NM_028039.2 | NP_082315.3 | ||
| AA189671.1 | ||||
| AK083728.1 | 1 | 3322 | ||
| AK144685.1 | BAE26012.1 | |||
| AK166457.1 | BAE38788.1 | |||
| BC033303.1 | AAH33303.2 | |||
| BC071275.1 | AAH71275.1 | |||
| CF730483.1 |
More Information
Set Membership: |
|

Genomic Map