Assay Details
Target Gene Details
| Entrez Gene ID: | 114741 | 
| Gene Name: | suppressor of Ty 16 | 
| Gene Aliases: | Cdc68, Fact140, Spt16, Supt16h | 
| Location: | Chr.14:52160419-52197239 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 2 - Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Supt16 | NM_033618.3 | NP_291096.2 | ||
| AF323667.1 | AAL04452.1 | |||
| AI930189.1 | ||||
| AK146529.1 | ||||
| AK148506.1 | BAE28591.1 | |||
| AK158116.1 | BAE34362.1 | |||
| AK169561.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top