Assay Details
Target Gene Details
Entrez Gene ID: | 69612 |
Gene Name: | KAT8 regulatory NSL complex subunit 2 |
Gene Aliases: |
2310037I24Rik, B930009D17 |
Location: |
Chr.15:98517658-98534732 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kansl2 | NM_001289437.1 | NP_001276366.1 | ||
| NM_001289438.1 | NP_001276367.1 | |||
| NM_001289439.1 | NP_001276368.1 | |||
| NM_001289440.1 | NP_001276369.1 | |||
| NM_133714.5 | NP_598475.2 | |||
| AK046649.1 | BAC32821.1 | |||
| AK046983.1 | BAC32935.1 | |||
| AK047269.1 | BAC33009.1 | |||
| AK089025.1 | ||||
| AK150748.1 | ||||
| AK151604.1 | ||||
| AK152604.1 | ||||
| AK153007.1 | ||||
| AK162714.1 | ||||
| AK167429.1 | BAE39517.1 | |||
| AK168499.1 | ||||
| AK183493.1 | ||||
| AK188822.1 | ||||
| AK205712.1 | ||||
| AK218567.1 | ||||
| BC003301.1 | AAH03301.1 | |||
| BC083325.1 | AAH83325.1 | |||
| BC092275.1 | AAH92275.1 | |||
| BC125631.1 | ||||
| BC125633.1 |

Genomic Map