Assay Details
Target Gene Details
Entrez Gene ID: | 69612 |
Gene Name: | KAT8 regulatory NSL complex subunit 2 |
Gene Aliases: |
2310037I24Rik, B930009D17 |
Location: |
Chr.15:98517658-98534732 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kansl2 | NM_001289437.1 | NP_001276366.1 | ||
NM_001289438.1 | NP_001276367.1 | |||
NM_001289439.1 | NP_001276368.1 | |||
NM_001289440.1 | NP_001276369.1 | |||
NM_133714.5 | NP_598475.2 | |||
AK009662.1 | BAC25264.1 | |||
AK046649.1 | BAC32821.1 | |||
AK046983.1 | BAC32935.1 | |||
AK047269.1 | BAC33009.1 | |||
AK089025.1 | ||||
AK150748.1 | ||||
AK151604.1 | ||||
AK152604.1 | ||||
AK153007.1 | ||||
AK162714.1 | ||||
AK167429.1 | BAE39517.1 | |||
AK168499.1 | ||||
BC003301.1 | AAH03301.1 | |||
BC083325.1 | AAH83325.1 | |||
BC092275.1 | AAH92275.1 | |||
BC125631.1 | ||||
BC125633.1 |
