Assay Details
Target Gene Details
Entrez Gene ID: | 114741 |
Gene Name: | suppressor of Ty 16 |
Gene Aliases: |
Cdc68, Fact140, Spt16, Supt16h |
Location: |
Chr.14:52160419-52197239 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt16 | NM_033618.3 | 5 | 652 | NP_291096.2 |
| AF323667.1 | 5 | 614 | AAL04452.1 | |
| AI930189.1 | 4 | 335 | ||
| AK146529.1 | 5 | 632 | ||
| AK148506.1 | 5 | 605 | BAE28591.1 | |
| AK158116.1 | 5 | 581 | BAE34362.1 | |
| AK169561.1 | 5 | 654 |
More Information
Set Membership: |
|

Genomic Map