Assay Details
Target Gene Details
Entrez Gene ID: | 94352 |
Gene Name: | lysyl oxidase-like 2 |
Gene Aliases: |
1110004B06Rik, 4930526G11Rik, 9430067E15Rik |
Location: |
Chr.14:69608692-69695834 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Loxl2 | NM_033325.2 | NP_201582.2 | ||
| AK034957.1 | BAC28894.1 | |||
| AK045019.1 | BAC32186.1 | |||
| AK145323.1 | BAE26367.1 | |||
| AK159386.1 | BAE35041.1 |
Target Gene Details
Entrez Gene ID: | 102465573 |
Gene Name: | microRNA 6950 |
Gene Aliases: |
Gm27586, mmu-mir-6950 |
Location: |
Chr.14:69692248-69692320 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6950 | NR_105915.1 |
More Information
Set Membership: |
|

Genomic Map