Assay Details
Target Gene Details
Entrez Gene ID: | 110265 |
Gene Name: | methionine sulfoxide reductase A |
Gene Aliases: |
2310045J23Rik, 6530413P12Rik, MSR-A |
Location: |
Chr.14:64122621-64455903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msra | NM_026322.4 | NP_080598.2 | ||
| BC089311.1 | AAH89311.1 |
More Information
Set Membership: |
|

Genomic Map