Assay Details
Target Gene Details
Entrez Gene ID: | 52033 |
Gene Name: | PDZ binding kinase |
Gene Aliases: |
2810434B10Rik, AW538537, D14Ertd732e, TOPK |
Location: |
Chr.14:65805911-65817822 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pbk | NM_023209.2 | NP_075698.1 | ||
| AB041882.1 | BAA99578.1 | |||
| AK003838.1 | BAB23029.1 | |||
| AK076121.1 | 1 | 100 | BAC36199.1 | |
| BC020099.1 | 1 | 33 | AAH20099.1 |
More Information
Set Membership: |
|

Genomic Map