Assay Details
Target Gene Details
Entrez Gene ID: | 319508 |
Gene Name: | synaptotagmin XV |
Gene Aliases: |
CHR10SYT, E230025K04Rik, sytXV |
Location: |
Chr.14:34220046-34230421 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syt15 | NM_176931.2 | NP_795905.2 | ||
| NM_181529.4 | NP_852682.1 | |||
| AK042641.1 | ||||
| AK054180.1 | BAC35683.1 | |||
| BB639494.1 | ||||
| BC139313.1 | ||||
| BC139314.1 |
More Information
Set Membership: |
|

Genomic Map