Assay Details
Target Gene Details
Entrez Gene ID: | 319508 |
Gene Name: | synaptotagmin XV |
Gene Aliases: |
CHR10SYT, E230025K04Rik, sytXV |
Location: |
Chr.14:34220046-34230421 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syt15 | NM_176931.2 | 5 | 815 | NP_795905.2 |
| NM_181529.4 | 5 | 815 | NP_852682.1 | |
| AB109019.1 | 4 | 687 | BAC76814.1 | |
| AB109020.1 | 4 | 687 | BAC76815.1 | |
| AK042641.1 | 5 | 816 | ||
| AK054180.1 | 5 | 815 | BAC35683.1 | |
| BC139313.1 | 5 | 726 | ||
| BC139314.1 | 5 | 727 |
More Information
Set Membership: |
|

Genomic Map