Assay Details
Target Gene Details
Entrez Gene ID: | 71978 |
Gene Name: | protein phosphatase 2, regulatory subunit B, alpha |
Gene Aliases: |
2410004D02Rik, mKIAA1541 |
Location: |
Chr.14:67014056-67072471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r2a | NM_001205188.1 | NP_001192117.1 | ||
| NM_028032.3 | NP_082308.1 | |||
| AK010380.1 | BAB26898.1 | |||
| AK090027.1 | ||||
| AK161507.1 | BAE36431.1 | |||
| AK161704.1 | BAE36540.1 | |||
| AK164497.1 | BAE37812.1 | |||
| AK167189.1 | BAE39320.1 | |||
| AK167530.1 | ||||
| AK184217.1 | ||||
| AK220192.1 | BAD90377.1 | |||
| BC065100.1 | AAH65100.1 | |||
| BC080692.1 | AAH80692.1 |
More Information
Set Membership: |
|

Genomic Map