Assay Details
Target Gene Details
Entrez Gene ID: | 75602 |
Gene Name: | RIKEN cDNA 1810062O18 gene |
Gene Aliases: |
- |
Location: |
Chr.14:20546293-20570680 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810062O18Rik | NR_033571.1 | |||
| AK007938.1 | BAB25363.1 |
Target Gene Details
Entrez Gene ID: | 19056 |
Gene Name: | protein phosphatase 3, catalytic subunit, beta isoform |
Gene Aliases: |
1110063J16Rik, Calnb, CnAbeta, Cnab |
Location: |
Chr.14:20499314-20546573 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp3cb | NM_008914.2 | NP_032940.1 | ||
| AB221678.1 | ||||
| AK135203.1 | BAE22459.1 | |||
| BC066000.1 | AAH66000.1 | |||
| M81483.1 | AAA37411.1 |
More Information
Set Membership: |
|

Genomic Map