Assay Details
Target Gene Details
Entrez Gene ID: | 19229 |
Gene Name: | PTK2 protein tyrosine kinase 2 beta |
Gene Aliases: |
CADTK, CAKB, CAKbeta, E430023O05Rik, FADK2, FAK2, PYK2, Raftk |
Location: |
Chr.14:66153257-66281066 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptk2b | NM_001162365.1 | NP_001155837.1 | ||
| NM_001162366.1 | NP_001155838.1 | |||
| NM_172498.3 | NP_766086.2 | |||
| AA462640.1 | ||||
| AI181574.1 | ||||
| AK088696.1 | BAC40510.1 | |||
| AK150105.1 | BAE29312.1 | |||
| BC137704.1 | ||||
| BC144849.1 | ||||
| JN971017.1 |

Genomic Map