Assay Details
Target Gene Details
Entrez Gene ID: | 67296 |
Gene Name: | suppressor of cytokine signaling 4 |
Gene Aliases: |
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7 |
Location: |
Chr.14:47277143-47291591 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs4 | NM_080843.2 | 1 | 37 | NP_543119.2 |
| AK014115.1 | 1 | 69 | BAC25425.1 | |
| BC057159.1 | 1 | 168 | AAH57159.1 |
More Information
Set Membership: |
|

Genomic Map