Assay Details
Target Gene Details
Entrez Gene ID: | 18753 |
Gene Name: | protein kinase C, delta |
Gene Aliases: |
AI385711, D14Ertd420e, PKC[d], PKCdelta, Pkcd |
Location: |
Chr.14:30595353-30626414 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkcd | NM_011103.3 | 10 | 1170 | NP_035233.1 |
| AB011812.1 | 9 | 933 | BAA36408.1 | |
| AB201453.1 | 7 | 772 | ||
| AB201454.1 | 7 | 772 | ||
| AB201455.1 | 7 | 772 | ||
| AB201456.1 | 7 | 772 | ||
| AF251036.1 | 9 | 933 | AAF64316.1 | |
| AK144231.1 | 10 | 1175 | BAE25785.1 | |
| AK168768.1 | 10 | 1165 | BAE40604.1 | |
| AK171476.1 | 10 | 1172 | ||
| AY545076.1 | 9 | 933 | AAS57795.1 | |
| BC051416.1 | 10 | 1134 | AAH51416.1 | |
| HQ840432.1 | ||||
| M69042.1 | 10 | 1165 | AAA73056.1 | |
| X60304.1 | 9 | 946 | CAA42845.1 |
More Information
Set Membership: |
|

Genomic Map