Assay Details
Target Gene Details
Entrez Gene ID: | 19270 |
Gene Name: | protein tyrosine phosphatase, receptor type, G |
Gene Aliases: |
5430405N12Rik, AW046354, AW549872, RPTPgamma |
Location: |
Chr.14:11553553-12242039 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprg | NM_008981.3 | NP_033007.2 | ||
| AK144283.1 | BAE25813.1 | |||
| L09562.1 | AAA40022.1 |
More Information
Set Membership: |
|

Genomic Map