Assay Details
Target Gene Details
Entrez Gene ID: | 54650 |
Gene Name: | Scm-like with four mbt domains 1 |
Gene Aliases: |
4930442N21Rik, 9330180L21Rik, AA536974, Sfmbt, Smr |
Location: |
Chr.14:30714473-30822721 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfmbt1 | NM_001166531.1 | NP_001160003.1 | ||
| NM_001166532.1 | NP_001160004.1 | |||
| NM_019460.2 | NP_062333.1 | |||
| AB032165.1 | BAA96305.1 | |||
| AK142330.1 | ||||
| AK154826.1 | BAE32857.1 | |||
| AK160573.1 | BAE35882.1 | |||
| BC018217.1 | ||||
| BC036972.1 | AAH36972.1 | |||
| BC066188.1 | AAH66188.1 |
More Information
Set Membership: |
|

Genomic Map