Assay Details
Target Gene Details
Entrez Gene ID: | 218772 |
Gene Name: | retinoic acid receptor, beta |
Gene Aliases: |
A830025K23, Hap, Nr1b2 |
Location: |
Chr.14:16430839-17082331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rarb | NM_001289760.1 | NP_001276689.1 | ||
| NM_001289761.1 | NP_001276690.1 | |||
| NM_001289762.1 | NP_001276691.1 | |||
| NM_011243.2 | NP_035373.1 | |||
| AK043726.1 | ||||
| AK078471.1 | ||||
| BC076597.1 | AAH76597.1 | |||
| S56660.1 | AAB25784.2 |
More Information
Set Membership: |
|

Genomic Map