Assay Details
Target Gene Details
Entrez Gene ID: | 76355 |
Gene Name: | TDP-glucose 4,6-dehydratase |
Gene Aliases: |
2610017J16Rik, 2610025M23Rik, AI648925 |
Location: |
Chr.14:118111911-118132765 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgds | NM_029578.3 | 9 | 785 | NP_083854.3 |
| AK011439.1 | 8 | 673 | ||
| AK011555.1 | 9 | 776 | BAB27693.1 | |
| AK154341.1 | 9 | 762 | BAE32527.1 | |
| AK165685.1 | 8 | 668 | ||
| AK165955.1 | 9 | 769 | BAE38483.1 | |
| BC021419.1 | 9 | 761 | AAH21419.1 |
More Information
Set Membership: |
|

Genomic Map