Assay Details
Target Gene Details
Entrez Gene ID: | 66480 |
Gene Name: | ribosomal protein L15 |
Gene Aliases: |
2510008H07Rik |
Location: |
Chr.14:18267823-18271024 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl15 | NM_025586.3 | NP_079862.1 | ||
| AK002234.1 | BAB21952.1 | |||
| AK010319.1 | BAB26850.1 | |||
| AK010674.1 | BAB27107.1 | |||
| AK010916.1 | BAB27266.1 | |||
| AK010931.1 | BAB27275.1 | |||
| AK012025.1 | BAB27981.1 | |||
| AK012423.1 | ||||
| AK151138.1 | BAE30145.1 | |||
| AK152711.1 | BAE31437.1 | |||
| AK159183.1 | BAE34881.1 | |||
| AK159626.1 | BAE35240.1 | |||
| AK167996.1 | BAE39986.1 | |||
| AK168125.1 | BAE40094.1 | |||
| AK169662.1 | BAE41284.1 | |||
| BC079842.1 | AAH79842.1 | |||
| BC087917.1 | AAH87917.1 |
More Information
Set Membership: |
|

Genomic Map