Assay Details
Target Gene Details
Entrez Gene ID: | 192187 |
Gene Name: | stabilin 1 |
Gene Aliases: |
FEEL-1, FELE-1, MFEEL-1, MS-1, STAB-1, mKIAA0246 |
Location: |
Chr.14:31139017-31168652 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 52 - Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stab1 | NM_138672.2 | NP_619613.2 | ||
| AB052959.1 | ||||
| AF290914.2 | AAL91671.2 | |||
| BC031166.1 | AAH31166.1 |
More Information
Set Membership: |
|

Genomic Map