Assay Details
Target Gene Details
Entrez Gene ID: | 20088 |
Gene Name: | ribosomal protein S24 |
Gene Aliases: |
- |
Location: |
Chr.14:24490633-24496146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps24 | NM_011297.2 | NP_035427.2 | ||
| NM_207634.1 | NP_997517.1 | |||
| NM_207635.1 | NP_997518.1 | |||
| AK002495.1 | BAB22143.1 | |||
| AK005358.1 | BAB23973.1 | |||
| AK007776.1 | BAB25248.1 | |||
| AK008386.1 | BAB25640.1 | |||
| AK009048.1 | BAB26046.1 | |||
| AK010856.1 | BAB27225.1 | |||
| AK012537.1 | BAB28304.1 | |||
| AK018704.1 | BAB31355.1 | |||
| AK052480.1 | ||||
| AK135247.1 | ||||
| AK151111.1 | BAE30121.1 | |||
| AK167874.1 | BAE39888.1 | |||
| BC058817.1 | AAH58817.1 | |||
| BC081457.1 | AAH81457.1 | |||
| BC086882.1 | AAH86882.1 | |||
| BC091748.1 | AAH91748.1 | |||
| BY311117.1 | ||||
| X60289.1 | CAA42829.1 |
More Information
Set Membership: |
|

Genomic Map