Assay Details
Target Gene Details
Entrez Gene ID: | 70021 |
Gene Name: | 5'-nucleotidase domain containing 2 |
Gene Aliases: |
2510015F01Rik |
Location: |
Chr.14:31131058-31139124 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5dc2 | NM_027289.1 | NP_081565.1 | ||
| AK010966.1 | BAB27298.1 | |||
| BC011230.1 | AAH11230.1 |
Target Gene Details
Entrez Gene ID: | 192187 |
Gene Name: | stabilin 1 |
Gene Aliases: |
FEEL-1, FELE-1, MFEEL-1, MS-1, STAB-1, mKIAA0246 |
Location: |
Chr.14:31139017-31168652 on Build GRCm38 |
Assay Gene Location: | Within Exon 69 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stab1 | NM_138672.2 | 69 | 7942 | NP_619613.2 |
| AB052959.1 | 69 | 7863 | ||
| AF290914.2 | 69 | 7873 | AAL91671.2 | |
| BC022136.1 | 8 | 1179 | AAH22136.1 | |
| BC031166.1 | 32 | 3796 | AAH31166.1 |
More Information
Set Membership: |
|

Genomic Map