Assay Details
Target Gene Details
Entrez Gene ID: | 67381 |
Gene Name: | mediator complex subunit 4 |
Gene Aliases: |
2410046H15Rik, DRIP36, HSPC126, TRAP36, Vdirp, Vdrip |
Location: |
Chr.14:73510049-73518545 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med4 | NM_026119.3 | NP_080395.1 | ||
| AK010270.1 | BAB26809.1 | |||
| AK010686.1 | BAB27118.1 | |||
| AK078288.1 | BAC37206.1 | |||
| AK145636.1 | BAE26554.1 | |||
| BC024950.1 | AAH24950.1 |
More Information
Set Membership: |
|

Genomic Map