Assay Details
Target Gene Details
Entrez Gene ID: | 218885 |
Gene Name: | oxidoreductase NAD-binding domain containing 1 |
Gene Aliases: |
2410002F01Rik, AW492292, BC019806 |
Location: |
Chr.14:32085728-32103203 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxnad1 | NM_145460.2 | NP_663435.2 | ||
| AK028649.1 | ||||
| AK031978.1 | BAC27636.1 | |||
| AK032160.1 | ||||
| AK075924.1 | ||||
| AK139884.1 | BAE24171.1 | |||
| AK215115.1 | ||||
| AK215185.1 | ||||
| BC019806.1 | AAH19806.1 |
More Information
Set Membership: |
|

Genomic Map