Assay Details
Target Gene Details
Entrez Gene ID: | 545136 |
Gene Name: | family with sequence similarity 186, member B |
Gene Aliases: |
EG545136 |
Location: |
Chr.15:99271018-99295891 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam186b | NM_001081254.1 | 7 | 2678 | NP_001074723.1 |

Genomic Map