Assay Details
Target Gene Details
Entrez Gene ID: | 106014 |
Gene Name: | family with sequence similarity 19, member A5 |
Gene Aliases: |
TAFA5, Tafa-5, Tara-5 |
Location: |
Chr.15:87544299-87759364 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam19a5 | NM_001252310.1 | NP_001239239.1 | ||
| NM_134096.2 | NP_598857.1 | |||
| NR_045510.1 | ||||
| AK090194.1 | BAC41130.1 | |||
| AK090307.1 | ||||
| AY325124.1 | AAP92416.1 | |||
| BB873716.1 | ||||
| BC015306.1 | AAH15306.1 | |||
| BI986174.1 | ||||
| BU057253.1 | ||||
| BY733542.1 |
More Information
Set Membership: |
|

Genomic Map