Assay Details
Target Gene Details
Entrez Gene ID: | 76282 |
Gene Name: | glutamic pyruvic transaminase, soluble |
Gene Aliases: |
1300007J06Rik, 2310022B03Rik, ALT, Gpt-1, Gpt1 |
Location: |
Chr.15:76696764-76699675 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpt | NM_182805.2 | NP_877957.1 | ||
| AK004932.1 | 2 | 650 | ||
| AK008086.1 | ||||
| AK009457.1 | ||||
| BC022625.1 | AAH22625.1 | |||
| BC026846.1 | AAH26846.1 |
More Information
Set Membership: |
|

Genomic Map