Assay Details
Target Gene Details
Entrez Gene ID: | 11838 |
Gene Name: | activity regulated cytoskeletal-associated protein |
Gene Aliases: |
Arc3.1, C86064, arg3.1, mArc |
Location: |
Chr.15:74669081-74672570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arc | NM_001276684.1 | NP_001263613.1 | ||
| NM_018790.3 | NP_061260.1 | |||
| AF162777.1 | AAD43586.1 | |||
| AK157822.1 | BAE34212.1 | |||
| BC023127.1 | AAH23127.1 |

Genomic Map