Assay Details
Target Gene Details
Entrez Gene ID: | 76282 |
Gene Name: | glutamic pyruvic transaminase, soluble |
Gene Aliases: |
1300007J06Rik, 2310022B03Rik, ALT, Gpt-1, Gpt1 |
Location: |
Chr.15:76696764-76699675 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpt | NM_182805.2 | 9 | 1344 | NP_877957.1 |
| AK004932.1 | 2 | 2236 | ||
| AK008086.1 | 9 | 1367 | ||
| AK009457.1 | 9 | 1367 | ||
| BC022625.1 | 9 | 1345 | AAH22625.1 | |
| BC026846.1 | 9 | 1343 | AAH26846.1 |
More Information
Set Membership: |
|

Genomic Map