Assay Details
Target Gene Details
Entrez Gene ID: | 14114 |
Gene Name: | fibulin 1 |
Gene Aliases: |
- |
Location: |
Chr.15:85205944-85286294 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln1 | NM_010180.2 | 12 | 1513 | NP_034310.2 |
AK035388.1 | 12 | 1466 | BAC29054.1 | |
AK083573.1 | 12 | 1516 | ||
AK085989.1 | 12 | 1456 | ||
AK086451.1 | 12 | 1514 | BAC39669.1 | |
AK137511.1 | 12 | 1460 | ||
AK159646.1 | 12 | 1468 | BAE35258.1 | |
BC007140.1 | 12 | 1457 | AAH07140.1 | |
EU543224.1 | 12 | 1451 | ||
X70854.1 | 12 | 1510 | CAA50207.1 |
