Assay Details
Target Gene Details
Entrez Gene ID: | 14114 |
Gene Name: | fibulin 1 |
Gene Aliases: |
- |
Location: |
Chr.15:85205944-85286294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln1 | NM_010180.2 | NP_034310.2 | ||
AK085989.1 | ||||
AK137511.1 | ||||
AK159646.1 | BAE35258.1 | |||
AK159816.1 | BAE35396.1 | |||
EU543224.1 | ||||
X70854.1 | CAA50207.1 |
