Assay Details
Target Gene Details
Entrez Gene ID: | 11514 |
Gene Name: | adenylate cyclase 8 |
Gene Aliases: |
AC8, AW060868 |
Location: |
Chr.15:64699035-64922296 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adcy8 | NM_001291903.1 | 11 | 3729 | NP_001278832.1 |
NM_009623.2 | 12 | 3819 | NP_033753.2 | |
AK138244.1 | 11 | 3730 | BAE23593.1 | |
U85021.1 | 12 | 3679 | AAB41885.1 |
