Assay Details
Target Gene Details
Entrez Gene ID: | 11514 |
Gene Name: | adenylate cyclase 8 |
Gene Aliases: |
AC8, AW060868 |
Location: |
Chr.15:64699035-64922296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy8 | NM_001291903.1 | NP_001278832.1 | ||
| NM_009623.2 | NP_033753.2 | |||
| AK138244.1 | BAE23593.1 | |||
| AK163443.1 | ||||
| U85021.1 | AAB41885.1 |

Genomic Map